Learn More
Medchemexpress LLC HY-12340 5mg Medchemexpress, ETP-46321 CAS:1252594-99-2 Purity:>98%
SDP
Catalog No. 502081015
Encompass_Preferred
Medchemexpress, HY-12340 5mg ETP-46321 CAS:1252594-99-2 ETP-46321 is a potent and orally bioavailable PI3Kα and PI3Kδ inhibitor with Kiapps of 2.3 and 14.2 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1015 Supplier Medchemexpress LLC Supplier No. HY123405MG
Medchemexpress, HY-12340 5mg ETP-46321 CAS:1252594-99-2 ETP-46321 is a potent and orally bioavailable PI3Kα and PI3Kδ inhibitor with Kiapps of 2.3 and 14.2 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.