Learn More
Medchemexpress LLC HY-12789 5mg Medchemexpress, Etrasimod CAS:1206123-37-6 Purity:>98%
SDP
Catalog No. 502081142
Encompass_Preferred
Medchemexpress, HY-12789 5mg Etrasimod CAS:1206123-37-6 Etrasimod (APD334) is a potent, selective and orally available antagonist of the sphingosine-1-phosphate-1 (S1P1) receptor with an IC50 value of 1.88 nM in CHO cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1142 Supplier Medchemexpress LLC Supplier No. HY127895MG
Medchemexpress, HY-12789 5mg Etrasimod CAS:1206123-37-6 Etrasimod (APD334) is a potent, selective and orally available antagonist of the sphingosine-1-phosphate-1 (S1P1) receptor with an IC50 value of 1.88 nM in CHO cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.