Learn More
Medchemexpress LLC Etravirine-d4 | 1142095-93-9 | 98.3% | C20H11D4BrN6O | 1 MG
SDP
Catalog No. 5000432454
Encompass_Preferred
Etravirine-d4 is the deuterium-labeled version of Etravirine, a non-nucleoside reverse transcriptase inhibitor (NNRTI) used in the treatment of HIV. This compound is intended for research use only and is not sold to patients.
  • Can be utilized as a tracer
  • Can be used as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS
Catalog No. 50-004-32454 Supplier Medchemexpress LLC Supplier No. HY90005S1MG
Etravirine-d4 is the deuterium-labeled version of Etravirine, a non-nucleoside reverse transcriptase inhibitor (NNRTI) used in the treatment of HIV. This compound is intended for research use only and is not sold to patients.
  • Can be utilized as a tracer
  • Can be used as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS