Learn More
Apexbio Technology LLC Etravirine (TMC125) 269055-15-4 1g
SDP
Catalog No. 5000094152
Encompass_Preferred
Etravirine (TMC125) (CAS 269055-15-4) is a small-molecule inhibitor targeting HIV-1 reverse transcriptase It is designed to block the activity of this enzyme thereby preventing HIV-1 viral replication Etravirine exerts its biological activity primarily through direct binding to HIV-1 reverse transcriptase as a non-nucleoside reverse transcriptase inhibitor (NNRTI) inducing conformational changes that reduce the enzyme s catalytic efficiency In cell-based assays Etravirine demonstrates potent inhibition of HIV-1 replication with reported IC50 values typically in the low nanomolar to sub-nanomolar range Based on these pharmacological properties Etravirine holds research potential in antiviral activity profiling resistance mutation analysis and mechanistic studies of reverse transcriptase function and HIV replication pathways
Catalog No. 50-000-94152 Supplier Apexbio Technology LLC Supplier No. B22241000
Etravirine (TMC125) (CAS 269055-15-4) is a small-molecule inhibitor targeting HIV-1 reverse transcriptase It is designed to block the activity of this enzyme thereby preventing HIV-1 viral replication Etravirine exerts its biological activity primarily through direct binding to HIV-1 reverse transcriptase as a non-nucleoside reverse transcriptase inhibitor (NNRTI) inducing conformational changes that reduce the enzyme s catalytic efficiency In cell-based assays Etravirine demonstrates potent inhibition of HIV-1 replication with reported IC50 values typically in the low nanomolar to sub-nanomolar range Based on these pharmacological properties Etravirine holds research potential in antiviral activity profiling resistance mutation analysis and mechanistic studies of reverse transcriptase function and HIV replication pathways