Learn More
Selleck Chemical LLC Etretinate S4699-20mg
SDP
Catalog No. 503108178
Encompass_Preferred
Etretinate (Tegison Ethyl etrinoate Retinoid Etretinato Ro 10-9359) is an oral aromatic retinoid acid which is effective in psoriasis and other dermatological syndromes It activates retinoid receptors causing an induction of cell differentiation inhibition of cell proliferation and inhibition of tissue infiltration by inflammatory cells
Catalog No. 50-310-8178 Supplier Selleck Chemical LLC Supplier No. S469920MG
Etretinate (Tegison Ethyl etrinoate Retinoid Etretinato Ro 10-9359) is an oral aromatic retinoid acid which is effective in psoriasis and other dermatological syndromes It activates retinoid receptors causing an induction of cell differentiation inhibition of cell proliferation and inhibition of tissue infiltration by inflammatory cells