Learn More
Medchemexpress LLC Etripamil | 1593673-23-4 | 98.1% | 452.59 | 250 MG
SDP
Catalog No. 5000385460
Encompass_Preferred
Etripamil is a short-acting, L-type calcium-channel antagonist that inhibits calcium influx through slow calcium channels, thereby slowing AV node conduction and prolonging the AV node refractory period. It increases heart rate and decreases systolic blood pressure, and is used in the study of paroxysmal supraventricular tachycardia (PSVT).
  • Inhibits calcium influx through slow calcium channels
  • Slows AV node conduction
  • Prolongs AV node refractory period
  • Increases heart rate and decreases systolic blood pressure
  • Used in the study of paroxysmal supraventricular tachycardia
Catalog No. 50-003-85460 Supplier Medchemexpress LLC Supplier No. 250MGHY17611
Etripamil is a short-acting, L-type calcium-channel antagonist that inhibits calcium influx through slow calcium channels, thereby slowing AV node conduction and prolonging the AV node refractory period. It increases heart rate and decreases systolic blood pressure, and is used in the study of paroxysmal supraventricular tachycardia (PSVT).
  • Inhibits calcium influx through slow calcium channels
  • Slows AV node conduction
  • Prolongs AV node refractory period
  • Increases heart rate and decreases systolic blood pressure
  • Used in the study of paroxysmal supraventricular tachycardia