Learn More
Selleck Chemical LLC Etrumadenant AB928-2mg
SDP
Supplier:  Selleck Chemical LLC S96082MG
Encompass_Preferred
Etrumadenant (AB928, A2aR/A2bR antagonist-1) is a novel dual A2aR/A2bR antagonist with Kd of 1.4 nM and 2 nM for A2aR and A2bR, respectively.
Catalog No. 50-238-9797