Learn More
Cole-Parmer ETS-D5 Programmable Temperature Probe
Supplier:  Cole-Parmer 0467155
Encompass
IKA 3378000 ETS-D5 Programmable Temperature Probe, Sensor features optimized PID control with three modes of operation — ensuring perfect temperature control without overshooting the set temperature, even in the case of quick heating. Mode A is suitable for work with varying parameters and is safety temperature adjustable; Mode B is suitable for series operation under uniform conditions; Mode C is suitable for unsupervised operation.
Catalog No. NC0970572