Learn More
Aviva Systems Biology Corp ETS2 Antibody - middle region (ARP38592_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38592_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ETS2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ETS2. Alternate names: ETS2IT1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-56718