Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jade Scientific, Inc ETT ACTIVATOR 0.5M 20L
SDP

Catalog No. 50326208 Shop All Jade Scientific, Inc Products
Encompass_Preferred

Ett Activator 0.5m 20l. 75-05-8

Catalog No. 50-326-208 Supplier Jade Scientific, Inc Supplier No. BJBR72520
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Ett Activator 0.5m 20l. 75-05-8

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.