Learn More
Aviva Systems Biology Corp ETV1 Antibody - middle region (ARP38528_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38528_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ETV1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ETV1. Alternate names: DKFZp781L0674, ER81, MGC104699, MGC120533, MGC120534ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56689