Learn More
Aviva Systems Biology Corp ETV2 Antibody - N-terminal region (ARP42806_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42806_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ETV2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ETV2. Alternate names: ER71, ETSRP71, MGC129834, MGC129835ApplicationWB. Species reactivity: Human
Catalog No. 50-285-027