Learn More
Aviva Systems Biology Corp ETV3L Antibody - C-terminal region (ARP47498_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47498_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ETV3L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ETV3L. Alternate names: FLJ16478ApplicationWB. Species reactivity: Dog, Human, Pig, Rabbit
Catalog No. 50-014-59569