Learn More
Aviva Systems Biology Corp ETV6 Antibody - C-terminal region (P100807_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100807_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ETV6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human ETV6. Alternate names: TEL, TEL/ABLApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53954