Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ETZ (C3-CA-DTZ) | 2989379-61-3 | 99.8% | C34H28N6O6 | 50 MG
SDP

Catalog No. 5000353325
Encompass_Preferred

ETZ (C3-CA-DTZ) is a promising luciferase prosubstrate that can be activated in vivo by nonspecific esterase to enhance the brain delivery of luciferin. It is intended for research use only and not sold to patients. This compound appears as a solid with a light yellow to orange color.

  • Promising luciferase substrate (prosubstrate)
  • Activatable in vivo by nonspecific esterase
  • Enhances brain delivery of luciferin
  • For research use only
  • Solid appearance, light yellow to orange color
  • Molecular weight: 616.62
  • Shipped at room temperature in continental US
  • Store at -20°C, sealed, away from moisture and light (solid)
  • Store at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light (in solvent)

Catalog No. 50-003-53325 Supplier Medchemexpress LLC Supplier No. HY14512750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ETZ (C3-CA-DTZ) is a promising luciferase prosubstrate that can be activated in vivo by nonspecific esterase to enhance the brain delivery of luciferin. It is intended for research use only and not sold to patients. This compound appears as a solid with a light yellow to orange color.

  • Promising luciferase substrate (prosubstrate)
  • Activatable in vivo by nonspecific esterase
  • Enhances brain delivery of luciferin
  • For research use only
  • Solid appearance, light yellow to orange color
  • Molecular weight: 616.62
  • Shipped at room temperature in continental US
  • Store at -20°C, sealed, away from moisture and light (solid)
  • Store at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light (in solvent)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.