Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Etz (C3-CA-DTZ) | 2989379-61-3 | MFCD00072662 | 99.8% | C34H28N6O6 | 10MG
SDP

Catalog No. 5000259336
Encompass_Preferred

ETZ (C3-CA-DTZ) is a luciferase prosubstrate activatable in vivo by nonspecific esterases to enhance brain delivery of luciferin. It is supplied as a light yellow to orange solid with high purity and is soluble in DMSO, making it suitable for preparing concentrated stocks for in vivo bioluminescence experiments focused on brain imaging.

  • Activatable in vivo by nonspecific esterases to enhance brain delivery.
  • High purity suitable for sensitive assays.
  • Soluble in DMSO at ≥100 mg/mL for concentrated stock preparation.
  • Solid appearance facilitates handling and storage.
  • Storage-stable when sealed and protected from moisture and light.

Catalog No. 50-002-59336 Supplier Medchemexpress LLC Supplier No. HY14512710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ETZ (C3-CA-DTZ) is a luciferase prosubstrate activatable in vivo by nonspecific esterases to enhance brain delivery of luciferin. It is supplied as a light yellow to orange solid with high purity and is soluble in DMSO, making it suitable for preparing concentrated stocks for in vivo bioluminescence experiments focused on brain imaging.

  • Activatable in vivo by nonspecific esterases to enhance brain delivery.
  • High purity suitable for sensitive assays.
  • Soluble in DMSO at ≥100 mg/mL for concentrated stock preparation.
  • Solid appearance facilitates handling and storage.
  • Storage-stable when sealed and protected from moisture and light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.