Learn More
Molecular Devices Molecular Devices ScanLater Wash Buffer, 10X, 100 mL
Catalog No. 5000014653
Encompass_Preferred
Molecular Devices ScanLater Wash Buffer is a 10X concentrated washing solution supplied in a 100 mL bottle for use in ScanLater western blot workflows. It is formulated to remove unbound reagents and support reliable europium-based detection.
  • 10X concentrated wash solution for dilution to working strength.
  • 100 mL liquid format suitable for multiple membrane washes.
  • Formulated for efficient removal of unbound reagents and background reduction.
  • Compatible with europium-based detection chemistries used in time-resolved assays.
  • Provided with product insert containing dilution and use instructions.
Catalog No. 50-000-14653 Supplier Molecular Devices Supplier No. R8206
Molecular Devices ScanLater Wash Buffer is a 10X concentrated washing solution supplied in a 100 mL bottle for use in ScanLater western blot workflows. It is formulated to remove unbound reagents and support reliable europium-based detection.
  • 10X concentrated wash solution for dilution to working strength.
  • 100 mL liquid format suitable for multiple membrane washes.
  • Formulated for efficient removal of unbound reagents and background reduction.
  • Compatible with europium-based detection chemistries used in time-resolved assays.
  • Provided with product insert containing dilution and use instructions.