Learn More
Molecular Devices Molecular Devices Western Blot Assay Kit, Eu-Labeled Goat Anti-Rabbit, Evaluation
Catalog No. 5000014358
Encompass_Preferred
Eu-labeled goat anti-rabbit secondary antibody kit for sensitive, low-background time-resolved fluorescence detection of proteins on Western blot membranes. The kit supplies a europium-labeled goat anti-rabbit IgG and ScanLater buffers formulated for TRF imaging.
  • Eu-labeled goat anti-rabbit IgG, 20 μg.
  • 5X blocking buffer, 40 mL.
  • 10X washing buffer, 100 mL.
  • Enables time-resolved fluorescence detection with reduced background.
  • Store europium-labeled antibody at -20°C; buffers at 4°C.
  • For research use only; not for diagnostic procedures.
Catalog No. 50-000-14358 Supplier Molecular Devices Supplier No. R8202
Eu-labeled goat anti-rabbit secondary antibody kit for sensitive, low-background time-resolved fluorescence detection of proteins on Western blot membranes. The kit supplies a europium-labeled goat anti-rabbit IgG and ScanLater buffers formulated for TRF imaging.
  • Eu-labeled goat anti-rabbit IgG, 20 μg.
  • 5X blocking buffer, 40 mL.
  • 10X washing buffer, 100 mL.
  • Enables time-resolved fluorescence detection with reduced background.
  • Store europium-labeled antibody at -20°C; buffers at 4°C.
  • For research use only; not for diagnostic procedures.