Learn More
Molecular Devices Molecular Devices Western Blot Assay Kit, Eu-Labeled Streptavidin, Bulk, 60 μg
Catalog No. 5000014378
Encompass_Preferred
The ScanLater Eu-labeled Streptavidin Western Blot Assay Bulk Kit supplies reagents for fluorescent western blot membrane detection using compatible microplate reader detection cartridges. The kit includes Eu-labeled streptavidin, concentrated blocking buffer, and concentrated washing buffer sufficient for multiple mini gels. Intended for research use only.
  • eu-labeled streptavidin, 60 μg, lyophilized; reconstitute with 60 μL reagent-grade water.
  • 5X blocking buffer, 120 mL (1 bottle).
  • 10X washing buffer, 300 mL total (3 x 100 mL bottles).
  • recommended dilution for assay: 1:5000 for reconstituted eu-labeled streptavidin.
  • formulated for membrane detection with microplate reader cartridges; enables fluorescence-based western blot analysis.
  • for research use only; not for diagnostic procedures.
Catalog No. 50-000-14378 Supplier Molecular Devices Supplier No. R8203
The ScanLater Eu-labeled Streptavidin Western Blot Assay Bulk Kit supplies reagents for fluorescent western blot membrane detection using compatible microplate reader detection cartridges. The kit includes Eu-labeled streptavidin, concentrated blocking buffer, and concentrated washing buffer sufficient for multiple mini gels. Intended for research use only.
  • eu-labeled streptavidin, 60 μg, lyophilized; reconstitute with 60 μL reagent-grade water.
  • 5X blocking buffer, 120 mL (1 bottle).
  • 10X washing buffer, 300 mL total (3 x 100 mL bottles).
  • recommended dilution for assay: 1:5000 for reconstituted eu-labeled streptavidin.
  • formulated for membrane detection with microplate reader cartridges; enables fluorescence-based western blot analysis.
  • for research use only; not for diagnostic procedures.