Learn More
Molecular Devices Molecular Devices ScanLater Western Blot Kit, Eu-Labeled, 60 μg
Supplier:  Molecular Devices R8205
Encompass_Preferred
The ScanLater Eu-Labeled Goat Anti-Mouse Western Blot Assay Kit is a europium-based detection kit designed for sensitive, time-resolved fluorescence western blot analysis. It supplies Eu-labeled secondary antibody and the blocking and wash buffers needed for nitrocellulose or PVDF membrane detection.
  • Eu-labeled goat anti-mouse IgG, 60 μg.
  • Includes 5X blocking buffer, 120 mL.
  • Includes 10X washing buffer, 300 mL.
  • Intended for use with ScanLater time-resolved fluorescence detection workflows.
  • Antibody supplied lyophilized; reconstitute according to product insert.
Catalog No. 50-000-14489