Learn More
Molecular Devices Molecular Devices Western Blot Evaluation Kit, Europium-labeled, Goat Anti-Mouse
Catalog No. 5000014357
Encompass_Preferred
The ScanLater Anti-Mouse Evaluation Kit is a western blot detection evaluation kit containing a Europium-labeled goat anti-mouse antibody and supporting buffers for assay development. The kit provides reagents sufficient to evaluate multiple mini-gel western blots using the ScanLater detection workflow.
  • Europium-labeled goat anti-mouse antibody, 20 μg.
  • Includes blocking buffer, 40 mL.
  • Includes wash buffer, 100 mL.
  • Supports approximately 10 mini gels with the recommended protocol.
  • Provided as an evaluation/demo kit for assay optimization.
Catalog No. 50-000-14357 Supplier Molecular Devices Supplier No. R8201
The ScanLater Anti-Mouse Evaluation Kit is a western blot detection evaluation kit containing a Europium-labeled goat anti-mouse antibody and supporting buffers for assay development. The kit provides reagents sufficient to evaluate multiple mini-gel western blots using the ScanLater detection workflow.
  • Europium-labeled goat anti-mouse antibody, 20 μg.
  • Includes blocking buffer, 40 mL.
  • Includes wash buffer, 100 mL.
  • Supports approximately 10 mini gels with the recommended protocol.
  • Provided as an evaluation/demo kit for assay optimization.