Learn More
Beckman Coulter™ EUA IL-6 Calibrators S0 Through S5
Catalog No. 01912829 Shop All Beckman Coulter Products
Change view
Click to view available options
For Use With (Equipment):
Beckman Coulter Analyzers
Catalog No. For Use With (Equipment)
01-912-829 Beckman Coulter Analyzers
1 options
This item is not returnable. View return policy
Catalog No. 01-912-829 Supplier Beckman Coulter™ Supplier No. A30944
Add to Cart
Add to Cart
This item is not returnable. View return policy
Access IL-6 calibrators for use with Beckman Coulter analyzers.
  • For use with Beckman Coulter analyzers
  • The Access IL-6 Calibrators are intended to calibrate the Access IL-6 assay for the quantitative determination of IL-6 levels in human serum and plasma (heparin) using the Access Family of Immunoassay Systems.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Beckman Coulter Analyzers