Learn More
ATCC Mammaliicoccus sciuri; SC116
Catalog No. 502380673
Encompass
Mammaliicoccus sciuri strain SC116 is a bacterial type strain that was isolated from the skin of an eastern gray squirrel. This strain can be used in quality control testing.
Catalog No. 50-238-0673 Supplier ATCC Supplier No. 29062
Mammaliicoccus sciuri strain SC116 is a bacterial type strain that was isolated from the skin of an eastern gray squirrel. This strain can be used in quality control testing.