Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eucalyptol, Natural Isolate, Ricca Chemical
SDP

Catalog No. 01184923 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-184-923 20 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-184-923 Supplier Ricca Chemical Company Supplier No. RCCT906020C6

missing translation for 'validMainframeMsg'

Eucalyptol, Natural Isolate is a natural isolate food additive.

  • All Natural
  • Food Grade
  • FSSC 22000 and ISO 9001 Accreditated Facility
  • Additional Food Safety Testing Included (Pesticide, Residual Solvents, Heavy Metals)

Specifications

Chemical Name or Material Eucalyptol
Packaging Amber Glass
CAS 470-82-6
Certificate of Analysis Yes
Odor Camphoraceous, Pungent
Assay Percent Range 70% to 100%
Physical Form Liquid
Molecular Formula C10H18O
Color Colorless
Type Food Additive Standard
Molecular Weight (g/mol) 154.24
Quantity 20 g
Grade Food Grade
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.