Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eucalyptus Oil, FCC, 70%, Spectrum™ Chemical
SDP

Catalog No. 18602586 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-586 100 mL Amber Glass Bottle
18-602-587 500 mL Amber Glass Bottle
18-602-588 4 L Amber Glass Bottle
18-602-589 4 x 4 L Amber Glass Bottle
18-602-590 20 L Steel Can
18-602-591 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-602-586 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. EU106100ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

EU106, 8000-48-4

Spectrum™ Chemical Eucalyptus Oil, FCC is a fragarance and flavor additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8000-48-4

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Refractive Index 1.458-1.470
Quantity 100 mL
UN Number UN1197
Percent Purity 70%
Grade FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.