Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eucalyptus Oil, FCC, 70%, Spectrum™ Chemical
SDP

Catalog No. 18602591 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-591 200 L Amber Glass Bottle
18-602-586 100 mL Amber Glass Bottle
18-602-587 500 mL Amber Glass Bottle
18-602-588 4 L Amber Glass Bottle
18-602-589 4 x 4 L Amber Glass Bottle
18-602-590 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-602-591 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. EU106200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

EU106, 8000-48-4

Spectrum™ Chemical Eucalyptus Oil, FCC is a fragarance and flavor additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8000-48-4

Specifications

CAS Max % 100%
Boiling Point 175°C/347°F
Packaging Amber Glass Bottle
Refractive Index 1.458-1.470
Quantity 200 L
UN Number UN1197
Percent Purity 70%
Grade FCC
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Eucalyptus Oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.