Learn More
Medchemexpress LLC β-Eudesmol | 473-15-4 | 1 MG
SDP
Catalog No. 5000421659
Encompass_Preferred
β-Eudesmol is a sesquiterpene-like compound extracted from the rhizome of Atractylodes lancea. It exhibits anticancer and anti-inflammatory activities, induces apoptosis, and acts as a neostigmine antagonist, making it suitable for various biological studies.
  • Inhibits cell proliferation in various tumor cells
  • Reduces angiogenesis by inhibiting VEGF and bFGF induced proliferation in HUVEC cells
  • Induces apoptosis through caspase-3 activation
  • Acts as a neostigmine antagonist, preventing neuromuscular failure
  • Inhibits NF-κB signaling pathway, providing anti-inflammatory and antioxidant effects
  • Useful for sepsis disease research
Catalog No. 50-004-21659 Supplier Medchemexpress LLC Supplier No. HYN60181MG
β-Eudesmol is a sesquiterpene-like compound extracted from the rhizome of Atractylodes lancea. It exhibits anticancer and anti-inflammatory activities, induces apoptosis, and acts as a neostigmine antagonist, making it suitable for various biological studies.
  • Inhibits cell proliferation in various tumor cells
  • Reduces angiogenesis by inhibiting VEGF and bFGF induced proliferation in HUVEC cells
  • Induces apoptosis through caspase-3 activation
  • Acts as a neostigmine antagonist, preventing neuromuscular failure
  • Inhibits NF-κB signaling pathway, providing anti-inflammatory and antioxidant effects
  • Useful for sepsis disease research