Learn More
Medchemexpress LLC (-)-α-Eudesmol | 473-16-5 | 94.0% | 222.37 | 500 UG
SDP
Catalog No. 5000391266
Encompass_Preferred
(-)-α-Eudesmol is a diterpene dialdehyde that can be isolated from *Porella*. It has a protective effect against acute brain injury induced by diethylnitrosamine and is for research use only.
  • Diterpene dialdehyde
  • Isolated from *Porella*
  • Protective effect against acute brain injury
  • For research use only
Catalog No. 50-003-91266 Supplier Medchemexpress LLC Supplier No. HY165292500UG
(-)-α-Eudesmol is a diterpene dialdehyde that can be isolated from *Porella*. It has a protective effect against acute brain injury induced by diethylnitrosamine and is for research use only.
  • Diterpene dialdehyde
  • Isolated from *Porella*
  • Protective effect against acute brain injury
  • For research use only