Learn More
Apexbio Technology LLC Eugenol 97-53-0 100mg
SDP
Catalog No. 502240539
Encompass_Preferred
Eugenol (97-53-0) is a small-molecule modulator that exerts its biological activity by modulating intracellular glutathione (GSH) levels interrupting gap-junctional intercellular communication (GJIC) and inducing reactive oxygen species (ROS) It disrupts mitochondrial membrane potential (MMP) at concentrations between 10 and 100 M and at higher doses affects intracellular calcium homeostasis and pH stability In experimental cell-based studies eugenol demonstrates moderate MMP disruption with a reported cellular IC50 range of approximately 10 100 M depending on the context and cell type Based on these pharmacological properties eugenol holds research potential in the mitigation of oxidative stress gastric ulcer prevention and dental endodontic applications
This item is not returnable. View return policy
Catalog No. 50-224-0539 Supplier Apexbio Technology LLC Supplier No. B6149100
Eugenol (97-53-0) is a small-molecule modulator that exerts its biological activity by modulating intracellular glutathione (GSH) levels interrupting gap-junctional intercellular communication (GJIC) and inducing reactive oxygen species (ROS) It disrupts mitochondrial membrane potential (MMP) at concentrations between 10 and 100 M and at higher doses affects intracellular calcium homeostasis and pH stability In experimental cell-based studies eugenol demonstrates moderate MMP disruption with a reported cellular IC50 range of approximately 10 100 M depending on the context and cell type Based on these pharmacological properties eugenol holds research potential in the mitigation of oxidative stress gastric ulcer prevention and dental endodontic applications