Learn More
Selleck Chemical LLC Eugenol S4706-20mg
SDP
Catalog No. 503108187
Encompass_Preferred
Eugenol (4-Allyl-2-methoxyphenol 4-Allylguaiacol Eugenic acid Allylguaiacol) is a naturally occuring scent chemical found in clove oil and other plants The physiologic effect of eugenol is by means of increased Histamine Release and cell-mediated Immunity
Catalog No. 50-310-8187 Supplier Selleck Chemical LLC Supplier No. S470620MG
Add to Cart
Add to Cart
Eugenol (4-Allyl-2-methoxyphenol 4-Allylguaiacol Eugenic acid Allylguaiacol) is a naturally occuring scent chemical found in clove oil and other plants The physiologic effect of eugenol is by means of increased Histamine Release and cell-mediated Immunity