Learn More
Medchemexpress LLC Eugenol acetate | 93-28-7 | MFCD00026191 | 99.8% | 206.24 | C12H14O3 | 250 MG
SDP
Catalog No. 5000303622
Encompass_Preferred
Eugenol acetate (acetyleugenol) is a high-purity analytical reference standard supplied for research and analytical applications. It is a colorless to light-yellow solid/liquid mixture with molecular formula C12H14O3 and molecular weight 206.24 g/mol. Typical uses include chromatographic and spectrometric method development, compound identification, and biological research.
  • High purity suitable for analytical testing and reference use.
  • Available in multiple pack sizes, including 250 mg.
  • Provided with certificate of analysis and safety data sheet.
  • Suitable for chromatographic and spectrometric method development.
  • Useful for compound identification and biological assays.
Catalog No. 50-003-03622 Supplier Medchemexpress LLC Supplier No. 250MGHYW014612R
Eugenol acetate (acetyleugenol) is a high-purity analytical reference standard supplied for research and analytical applications. It is a colorless to light-yellow solid/liquid mixture with molecular formula C12H14O3 and molecular weight 206.24 g/mol. Typical uses include chromatographic and spectrometric method development, compound identification, and biological research.
  • High purity suitable for analytical testing and reference use.
  • Available in multiple pack sizes, including 250 mg.
  • Provided with certificate of analysis and safety data sheet.
  • Suitable for chromatographic and spectrometric method development.
  • Useful for compound identification and biological assays.