Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eugenol acetate | 93-28-7 | MFCD00026191 | 99.8% | 206.24 g/mol | C12H14O3 | 25 MG
SDP

Catalog No. 5000303415
Encompass_Preferred

Eugenol acetate is a high-purity analytical standard (acetyleugenol) intended for research and analytical use. It is supplied as a colorless to light yellow solid-liquid mixture for method development, quality control, and biochemical or pharmacological studies.

  • High purity: 99.8%.
  • Molecular formula C12H14O3.
  • Molecular weight 206.24 g/mol.
  • Supplied in small pack sizes suitable for analytical use.
  • Intended for method development, quality control, and reference standards.
  • Colorless to light yellow solid-liquid mixture.

Catalog No. 50-003-03415 Supplier Medchemexpress LLC Supplier No. 25MGHYW014612R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eugenol acetate is a high-purity analytical standard (acetyleugenol) intended for research and analytical use. It is supplied as a colorless to light yellow solid-liquid mixture for method development, quality control, and biochemical or pharmacological studies.

  • High purity: 99.8%.
  • Molecular formula C12H14O3.
  • Molecular weight 206.24 g/mol.
  • Supplied in small pack sizes suitable for analytical use.
  • Intended for method development, quality control, and reference standards.
  • Colorless to light yellow solid-liquid mixture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.