Learn More
Medchemexpress LLC Eugenol acetate | 93-28-7 | MFCD00026191 | 99.8% | 206.24 g/mol | C12H14O3 | 50 MG
SDP
Catalog No. 5000302750
Encompass_Preferred
Eugenol acetate (acetyleugenol) is an analytical standard and research reagent used in biochemical and analytical applications. It exhibits antibacterial, anti-inflammatory, antioxidant, and anticancer activity in vitro, and has been reported to inhibit NF-κB signaling while modulating p53 and p21 expression.
  • Analytical standard for research and quality control.
  • Reported antibacterial and antioxidant activity.
  • Inhibits NF-κB signaling in vitro.
  • Modulates p53 and p21 (WAF1) expression.
  • High purity listed at 99.83%.
  • Molecular formula C12H14O3; molecular weight 206.24 g/mol.
Catalog No. 50-003-02750 Supplier Medchemexpress LLC Supplier No. HYW014612R50MG
Eugenol acetate (acetyleugenol) is an analytical standard and research reagent used in biochemical and analytical applications. It exhibits antibacterial, anti-inflammatory, antioxidant, and anticancer activity in vitro, and has been reported to inhibit NF-κB signaling while modulating p53 and p21 expression.
  • Analytical standard for research and quality control.
  • Reported antibacterial and antioxidant activity.
  • Inhibits NF-κB signaling in vitro.
  • Modulates p53 and p21 (WAF1) expression.
  • High purity listed at 99.83%.
  • Molecular formula C12H14O3; molecular weight 206.24 g/mol.