Learn More
Medchemexpress LLC Eugenol-d3 | 1335401-17-6 | 98.5% | 167.22 | 1 MG
SDP
Catalog No. 5000427609
Encompass_Preferred
Eugenol-d3 is deuterium-labeled Eugenol. Eugenol is an essential oil found in cloves that exhibits antibacterial, anthelmintic, and antioxidant activities, and is shown to inhibit lipid peroxidation. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-27609 Supplier Medchemexpress LLC Supplier No. HYN0337S1MG
Eugenol-d3 is deuterium-labeled Eugenol. Eugenol is an essential oil found in cloves that exhibits antibacterial, anthelmintic, and antioxidant activities, and is shown to inhibit lipid peroxidation. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.