Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eugenol, USP, Spectrum™ Chemical
SDP

Catalog No. 18602593 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-593 500 mL Amber Glass Bottle
18-602-592 100 mL Amber Glass Bottle
18-602-594 4 L Amber Glass Bottle
18-602-595 20 L Amber Glass Bottle
18-602-596 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-602-593 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. EU110500MLGL
Only null left
Add to Cart
Add to Cart

EU110, 97-53-0, C10H12O2

Spectrum™ Chemical Eugenol, USP is used as a local antiseptic and anaesthetic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 97-53-0
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00008654
InChI Key RRAFCDWBNXTKKO-UHFFFAOYSA-N
IUPAC Name 2-methoxy-4-(prop-2-en-1-yl)phenol
SMILES COC1=CC(CC=C)=CC=C1O

Specifications

CAS Max % 100%
Boiling Point 253°C
Packaging Amber Glass Bottle
Refractive Index 1.540-1.542
Quantity 500 mL
Grade USP
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.