Learn More
Cayman Chemical EUK 118, Purity: ≥95%, CAS: 186299-34-3, 10 mg
SDP
Catalog No. 502889467
Encompass_Preferred
A synthetic catalytic scavenger of reactive oxygen species, with reduced activity compared to EUK 8 and EUK 134
Catalog No. 50-288-9467 Supplier Cayman Chemical Supplier No. 1027110 MG
A synthetic catalytic scavenger of reactive oxygen species, with reduced activity compared to EUK 8 and EUK 134