Learn More
Selleck Chemical LLC EUK 134 S4261-20mg
SDP
Catalog No. 503108194
Encompass_Preferred
EUK 134 a synthetic superoxide dismutase (SOD)/catalase mimetic exhibits potent antioxidant activities and inhibits the formation of -amyloid and related amyloid fibril
Catalog No. 50-310-8194 Supplier Selleck Chemical LLC Supplier No. S426120MG
EUK 134 a synthetic superoxide dismutase (SOD)/catalase mimetic exhibits potent antioxidant activities and inhibits the formation of -amyloid and related amyloid fibril