Learn More
Selleck Chemical LLC EUK 134 S4261-10mM/1mL
SDP
Catalog No. 503110458
Encompass_Preferred
EUK 134 a synthetic superoxide dismutase (SOD)/catalase mimetic exhibits potent antioxidant activities and inhibits the formation of -amyloid and related amyloid fibril
Catalog No. 50-311-0458 Supplier Selleck Chemical LLC Supplier No. S426110MM/1ML
EUK 134 a synthetic superoxide dismutase (SOD)/catalase mimetic exhibits potent antioxidant activities and inhibits the formation of -amyloid and related amyloid fibril