Learn More
Apexbio Technology LLC EUK 134, 20mg. Cas: 81065-76-1 MFCD: MFCD08059577. Salen-manganese complexes;SOD mimetic
SDP
Supplier:  Apexbio Technology LLC B132520
Encompass_Preferred
EUK 134, a synthetic superoxide dismutase (SOD)/catalase mimetic, has exhibited potent antioxidant activities and inhibited the formation of -amyloid and related amyloid fibril.In vitro:EUK-134, a salen-manganese complex, showed potent catalase and cytoprotective activities and SOD activity. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5216