Learn More
Apexbio Technology LLC EUK 134 81065-76-1 50mg
SDP
Supplier:  Apexbio Technology LLC B132550
Encompass_Preferred
EUK 134 (CAS 81065-76-1) is a synthetic salen-manganese complex designed to mimic the antioxidant enzymes superoxide dismutase (SOD) and catalase By scavenging reactive oxygen species (ROS) EUK 134 diminishes oxidative stress and attenuates cellular damage In vitro this compound inhibits formation of amyloid fibrils reduces microglial proliferation induced by -amyloid and protects neuronal cells from oxidative injury via inhibition of MAPK signaling In vivo studies indicate that EUK 134 decreases infarct size and neuronal cell death in rodent models of ischemic stroke and kainate-induced neurotoxicity EUK 134 is thus a relevant tool in oxidative stress and neurodegeneration research
Catalog No. 50-000-62063