Learn More
TARGETMOL CHEMICALS INC EUK-134 50MG
SDP
Catalog No. 502579348
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EUK-134 a synthetic superoxide dismutase (SOD)/catalase mimetic exhibits potent antioxidant activities and inhibits the formation of (beta)-amyloid and related amyloid fibril. purity: 99%
Catalog No. 50-257-9348 Supplier TARGETMOL CHEMICALS INC Supplier No. T649550MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EUK-134 a synthetic superoxide dismutase (SOD)/catalase mimetic exhibits potent antioxidant activities and inhibits the formation of (beta)-amyloid and related amyloid fibril. purity: 99%