Learn More
Medchemexpress LLC Euk-134 | 81065-76-1 | 98.3% | 416.74 g/mol | C18H18ClMnN2O4 | 10 MG
SDP
Catalog No. 5000213815
Encompass_Preferred
EUK-134 is a synthetic superoxide dismutase and catalase mimetic with mitoprotective antioxidant activity. It scavenges reactive oxygen species and has been reported to reduce markers of oxidative stress and protect against ischemia-reperfusion-induced tissue damage in cellular and animal models.
  • Acts as combined superoxide dismutase and catalase mimetic.
  • Reduces NF-κB expression and markers of lipid and protein oxidation.
  • Provides mitoprotective antioxidant effects in cellular models.
  • High purity solid form suitable for research use.
  • Available in small research quantities for assay development (10 mg).
  • Stable when stored protected from light under recommended temperatures.
Catalog No. 50-002-13815 Supplier Medchemexpress LLC Supplier No. HY10059410MG
EUK-134 is a synthetic superoxide dismutase and catalase mimetic with mitoprotective antioxidant activity. It scavenges reactive oxygen species and has been reported to reduce markers of oxidative stress and protect against ischemia-reperfusion-induced tissue damage in cellular and animal models.
  • Acts as combined superoxide dismutase and catalase mimetic.
  • Reduces NF-κB expression and markers of lipid and protein oxidation.
  • Provides mitoprotective antioxidant effects in cellular models.
  • High purity solid form suitable for research use.
  • Available in small research quantities for assay development (10 mg).
  • Stable when stored protected from light under recommended temperatures.