Learn More
Cayman Chemical EUK 134 Fatty Acids and Deriva
SDP
Catalog No. 502887939
Encompass_Preferred
A synthetic catalase/SOD mimic and hydrogen peroxide scavenger; consumes hydrogen peroxide with an initial reaction rate of 234 µM/min in vitro; reduces infarct volume >80% in rats at 2.5 mg/kg; attenuates brain damage in rats at 10 mg/kg following systemic administration of kainic acid
Catalog No. 50-288-7939 Supplier Cayman Chemical Supplier No. 10006329100 MG
A synthetic catalase/SOD mimic and hydrogen peroxide scavenger; consumes hydrogen peroxide with an initial reaction rate of 234 µM/min in vitro; reduces infarct volume >80% in rats at 2.5 mg/kg; attenuates brain damage in rats at 10 mg/kg following systemic administration of kainic acid