Learn More
Cayman Chemical EUK 134 Fatty Acids and Deriva
SDP
Supplier:  Cayman Chemical 100063295 MG
Encompass_Preferred
A synthetic catalase/SOD mimic and hydrogen peroxide scavenger; consumes hydrogen peroxide with an initial reaction rate of 234 µM/min in vitro; reduces infarct volume >80% in rats at 2.5 mg/kg; attenuates brain damage in rats at 10 mg/kg following systemic administration of kainic acid
Catalog No. 50-289-0802