Learn More
Enzo Life Sciences BIOARRAY™ Eukaryotic hybridization controls (150Reactions)
SDP
Catalog No. 502012527
Encompass_Preferred
BIOARRAY™ Eukaryotic Hybridization Controls contain biotin-labeled transcripts prepared from three E. coli genes, bioB, bioC and bioD, and one from bacteriophage P1, cre. These together with the synthetic biotin-labeled deoxyoligonucleotide control (Deoxyoligo B2) are added to biotin-labeled target cRNA before hybridization to probe sequences on the array. These sequences of known concentration provide an internal control for assaying quality and performance. Deoxyoligo B2 aids software programs such as the Affymetrix Microarray Suite by aligning the grid with the array image. Long Term Storage: -80°C.
Catalog No. 50-201-2527 Supplier Enzo Life Sciences Supplier No. ENZ42661150
BIOARRAY™ Eukaryotic Hybridization Controls contain biotin-labeled transcripts prepared from three E. coli genes, bioB, bioC and bioD, and one from bacteriophage P1, cre. These together with the synthetic biotin-labeled deoxyoligonucleotide control (Deoxyoligo B2) are added to biotin-labeled target cRNA before hybridization to probe sequences on the array. These sequences of known concentration provide an internal control for assaying quality and performance. Deoxyoligo B2 aids software programs such as the Affymetrix Microarray Suite by aligning the grid with the array image. Long Term Storage: -80°C.