Learn More
Apexbio Technology LLC eukaryotic translation elongation factor 1 alpha 1 (EEF1A1) (387-394) [Multiple species] 10mg
SDP
Supplier:  Apexbio Technology LLC A106710
Encompass_Preferred
eukaryotic translation elongation factor 1 alpha 1 EEF1A1 387-394 is a peptide fragment derived from EEF1A1 a protein centrally involved in protein synthesis by delivering aminoacyl-tRNAs to the ribosome This conserved peptide segment (LEU-GLU-ASP-GLY-PRO-LYS-PHE-LEU residues 387 394) is widely expressed across multiple species and serves as a reference antigen in biomedical research for studying autoimmune responses in certain diseases Based on these properties eukaryotic translation elongation factor 1 alpha 1 EEF1A1 387-394 is employed as a molecular tool to investigate translational regulation-associated cell death mechanisms and the immunological aspects of EEF1A1-related pathologies
Catalog No. 50-000-97040