Learn More
Electron Microscopy Sciences EUKITT Mounting Medium 100 ML
SDP
Catalog No. 50980467
Encompass_Preferred
Eukitt is a mounting medium (5% Acrylic Resin and 55% Xylenes) that is used in microscopy. It is an adhesive and specimen preservative that can be used manually and in automated coverslipping equipment. Special purpose adhesive and sealant for use with glass, non plastic and other non-reactive materials. It spreads quickly and evenly withoutforming any air bubbles. It is fast-drying (20 minutes) with a refractive index close to glass. Chemically neutral and has no reaction to staining.
Catalog No. 50-980-467 Supplier Electron Microscopy Sciences Supplier No. 15320
Eukitt is a mounting medium (5% Acrylic Resin and 55% Xylenes) that is used in microscopy. It is an adhesive and specimen preservative that can be used manually and in automated coverslipping equipment. Special purpose adhesive and sealant for use with glass, non plastic and other non-reactive materials. It spreads quickly and evenly withoutforming any air bubbles. It is fast-drying (20 minutes) with a refractive index close to glass. Chemically neutral and has no reaction to staining.