Learn More
Electron Microscopy Sciences EUKITT Mounting Medium, 500 mL
SDP
Catalog No. 50980468
Encompass_Preferred
The product holds up well in the presence of water and trace levels of Alcohol. It is very well suited for all pale color preparations as well as colorless preparations. EUKITT has a neutral pH and offers extended storage times of all slide preparations for years without any sign of cracking or discoloration. The medium itself is colorless and does not change the color and or the structure of the samples mounted with the medium. EUKITT is temperature resistant to heat and cold (-170°C), as well as humidity, daylight and UV light.
Catalog No. 50-980-468 Supplier Electron Microscopy Sciences Supplier No. 15322
The product holds up well in the presence of water and trace levels of Alcohol. It is very well suited for all pale color preparations as well as colorless preparations. EUKITT has a neutral pH and offers extended storage times of all slide preparations for years without any sign of cracking or discoloration. The medium itself is colorless and does not change the color and or the structure of the samples mounted with the medium. EUKITT is temperature resistant to heat and cold (-170°C), as well as humidity, daylight and UV light.