Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma-Aldrich Eukitt™ Quick-hardening Mounting Medium

Catalog No. 01259026 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01-259-026 100 mL
01-258-832 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-259-026 Supplier Sigma-Aldrich Supplier No. 03989100ML
Add to Cart
Add to Cart

Mounting medium for microscopy, containing 45% acrylic resin and 55% xylenes.

Eukitt™ Quick-hardening mounting medium, or Poly(butyl methacrylate-co-methyl methacrylate), is a mounting medium for microscopy, containing 45% acrylic resin and 55% xylenes. It is a special-purpose adhesive, sealant, and specimen preservative that can be used manually and in automated cover-slipping equipment, glass, non-plastic, and other non-reactive materials.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Slide Mounting Media
Reagent Type Mounting Solution
Volume (Metric) 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.