Learn More
Electron Microscopy Sciences EUKITT UV-R Low-Viscosity Mounting Medium 100 ML
SDP
Catalog No. 501929867
Encompass_Preferred
EUKITT UV mounting medium has all the well-known beneficial properties of the EUKITT mountants. EUKITT UV does not contain any solvents, therefore only 50% of the usual amount is needed compared to conventional systems. While using EUKITT UV almost no shrinkage can be detected.
The mounting medium may be used on tissue sections in non-polar solvents such as xylene or xylene substitutes as well as directly on sections in pure alcohols (EtOH, iPrOH, etc.).
The viscosity is very user-friendly. After being applied to the tissue, the cover glass can be placed as usual. Subsequently, a transfer break of at least 120 sec. is maintained prior to irradiation in order to ensure an ideal penetration into the tissue.
When using the specially tuned LED CCFL UV curing machine EUKITT UV cures in just 30 seconds.
Catalog No. 50-192-9867 Supplier Electron Microscopy Sciences Supplier No. 1532030R
EUKITT UV mounting medium has all the well-known beneficial properties of the EUKITT mountants. EUKITT UV does not contain any solvents, therefore only 50% of the usual amount is needed compared to conventional systems. While using EUKITT UV almost no shrinkage can be detected.
The mounting medium may be used on tissue sections in non-polar solvents such as xylene or xylene substitutes as well as directly on sections in pure alcohols (EtOH, iPrOH, etc.).
The viscosity is very user-friendly. After being applied to the tissue, the cover glass can be placed as usual. Subsequently, a transfer break of at least 120 sec. is maintained prior to irradiation in order to ensure an ideal penetration into the tissue.
When using the specially tuned LED CCFL UV curing machine EUKITT UV cures in just 30 seconds.